Product Details

SNP ID
rs1891511
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:115830468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCAGCTACCCCAGTACACCCC[A/G]CAGGGCAACTTTGGGCCATGGATAG
Phenotype
MIM: 162361
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NHLH2 PubMed Links

Gene Details

Gene
NHLH2
Gene Name
nescient helix-loop-helix 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111061.1 Intron NP_001104531.1
NM_005599.3 Intron NP_005590.1
XM_017001370.1 Intron XP_016856859.1

View Full Product Details