Product Details

SNP ID
rs6495306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78573551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAACGAATTGAACAAGATATCAC[A/G]TTAGGCTCTAGTGTGGCCAAGTTGT
Phenotype
MIM: 118505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNA5 PubMed Links

Gene Details

Gene
CHRNA5
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000745.3 Intron NP_000736.2
NM_001307945.1 Intron NP_001294874.1
XM_005254142.3 Intron XP_005254199.1
XM_017021881.1 Intron XP_016877370.1

View Full Product Details