Product Details
- SNP ID
-
rs3866543
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:76337268 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGGTTGTTGTTTGTTTGCCTTTCGG[G/T]TTTTCAAAGCTAGGAAGCTTTCTTT
- Phenotype
-
MIM: 609481
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ISL2
PubMed Links
Gene Details
- Gene
- ISL2
- Gene Name
- ISL LIM homeobox 2
View Full Product Details