Product Details

SNP ID
rs1982873
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:39955579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGCTGTGGGCGCCCTCGCCCC[C/T]CTTTGCTCACCCGCACTCCACGCTC
Phenotype
MIM: 614397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD2A PubMed Links

Gene Details

Gene
MFSD2A
Gene Name
major facilitator superfamily domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136493.2 Intron NP_001129965.1
NM_001287808.1 Intron NP_001274737.1
NM_001287809.1 Intron NP_001274738.1
NM_032793.4 Intron NP_116182.2
XM_005271285.1 Intron XP_005271342.1
XM_011542312.2 Intron XP_011540614.1

View Full Product Details