Product Details

SNP ID
rs1168007
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:62473963 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGTCTCAATTTGAAGATCTTTA[C/T]GCAGAAAGCCTTGAATACCTTTTAG
Phenotype
MIM: 615730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK7 PubMed Links

Gene Details

Gene
DOCK7
Gene Name
dedicator of cytokinesis 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271999.1 Intron NP_001258928.1
NM_001272000.1 Intron NP_001258929.1
NM_001272001.1 Intron NP_001258930.1
NM_001272002.1 Intron NP_001258931.1
NM_033407.3 Intron NP_212132.2
XM_005271292.2 Intron XP_005271349.1
XM_011542326.2 Intron XP_011540628.1
XM_011542327.2 Intron XP_011540629.1
XM_011542328.2 Intron XP_011540630.1
XM_011542330.2 Intron XP_011540632.1
XM_017002639.1 Intron XP_016858128.1
XM_017002640.1 Intron XP_016858129.1

View Full Product Details