Product Details
- SNP ID
-
rs1934697
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:80353117 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATGTTCCTCCTAAGCCATGAGAGC[C/T]GTTTTTACAAAGCATCACAATAGTC
- Phenotype
-
MIM: 615154
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DYDC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79192278] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DYDC1
- Gene Name
- DPY30 domain containing 1
- Gene
- DYDC2
- Gene Name
- DPY30 domain containing 2
View Full Product Details