Product Details

SNP ID
rs34767676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4145278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTTAAAATACAAGCCACTTCC[C/T]TCTGTCTAGGAAGTACTAAAGAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5D2 PubMed Links

Gene Details

Gene
CYB5D2
Gene Name
cytochrome b5 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254755.1 1186 Intron NP_001241684.1
NM_001254756.1 1186 Intron NP_001241685.1
NM_144611.3 1186 Intron NP_653212.1
XM_006721447.2 1186 UTR 5 XP_006721510.2
XM_011523649.2 1186 Intron XP_011521951.1
XM_017024180.1 1186 Intron XP_016879669.1
XM_017024181.1 1186 Intron XP_016879670.1
XM_017024182.1 1186 UTR 5 XP_016879671.1
XM_017024183.1 1186 Intron XP_016879672.1
Gene
ZZEF1
Gene Name
zinc finger ZZ-type and EF-hand domain containing 1
There are no transcripts associated with this gene.

View Full Product Details