Product Details
- SNP ID
-
rs14655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:86970577 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTGGGAAAAAATTCGGCCTCCTG[A/C]AATGACAGCAGGGAGACTTGGGTCG
- Phenotype
-
MIM: 608925
MIM: 602998
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADIRF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7960] are located under a probe and SNP(s) [rs74503710] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADIRF
- Gene Name
- adipogenesis regulatory factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006829.2 |
735 |
Missense Mutation |
AAA,CAA |
K76Q |
NP_006820.1 |
- Gene
- AGAP11
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_133447.1 |
735 |
Intron |
|
|
NP_597704.1 |
- Gene
- MMRN2
- Gene Name
- multimerin 2
- Gene
- SNCG
- Gene Name
- synuclein gamma
There are no transcripts associated with this gene.
View Full Product Details