Product Details

SNP ID
rs2296075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113317151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGCATACCATCACCAAGAAGTGT[C/T]GGGGTCTCACCGGCTAAAGACATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR31 PubMed Links

Gene Details

Gene
WDR31
Gene Name
WD repeat domain 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006615.2 Intron NP_001006616.1
NM_001012361.3 Intron NP_001012361.1
NM_145241.4 Intron NP_660284.1
XM_011518194.2 Intron XP_011516496.1

View Full Product Details