Product Details

SNP ID
rs2774620
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:17135623 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTCCCCAAACCCGGGCCCCTGCCC[C/T]TTTCCGAACTGCGGCCAGACAGGCT
Phenotype
MIM: 611870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTLN PubMed Links

Gene Details

Gene
CNTLN
Gene Name
centlein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114395.2 Intron NP_001107867.1
NM_001286984.1 Intron NP_001273913.1
NM_001286985.1 Intron NP_001273914.1
NM_017738.3 Intron NP_060208.2
XM_005251492.1 Intron XP_005251549.1
XM_006716793.3 Intron XP_006716856.1
XM_011517941.2 Intron XP_011516243.1
XM_017014839.1 Intron XP_016870328.1
XM_017014840.1 Intron XP_016870329.1
XM_017014841.1 Intron XP_016870330.1
XM_017014842.1 Intron XP_016870331.1
XM_017014843.1 Intron XP_016870332.1
XM_017014844.1 Intron XP_016870333.1
XM_017014845.1 Intron XP_016870334.1
XM_017014846.1 Intron XP_016870335.1
XM_017014847.1 Intron XP_016870336.1

View Full Product Details