Product Details

SNP ID
rs2296490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55152758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAGTTTAGGGAATGGATCAAAC[A/G]TGGAGCCTGGTGCAAGATCAGGCGT
Phenotype
MIM: 153619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLGAP5 PubMed Links

Gene Details

Gene
DLGAP5
Gene Name
DLG associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146015.1 Intron NP_001139487.1
NM_014750.4 Intron NP_055565.3
XM_017021840.1 Intron XP_016877329.1
Gene
LGALS3
Gene Name
lectin, galactoside binding soluble 3
There are no transcripts associated with this gene.

View Full Product Details