Product Details

SNP ID
rs2301151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35352183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGCTGCCTCCGGGCACTGGCCC[A/G]CTCCGGCCTGACGCACAGGCGGAAG
Phenotype
MIM: 107266 MIM: 603820 MIM: 603821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD22 PubMed Links

Gene Details

Gene
CD22
Gene Name
CD22 molecule
There are no transcripts associated with this gene.

Gene
FFAR1
Gene Name
free fatty acid receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005303.2 642 Missense Mutation CAC,CGC H211R NP_005294.1
Gene
FFAR3
Gene Name
free fatty acid receptor 3
There are no transcripts associated with this gene.

Gene
MIR5196
Gene Name
microRNA 5196
There are no transcripts associated with this gene.

View Full Product Details