Product Details

SNP ID
rs2301268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15429555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGGATGATGAGACAATTTTCCT[C/G]TCTCCCTTGGGAGAAGGGAATATCT
Phenotype
MIM: 608315
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EAF1 PubMed Links

Gene Details

Gene
EAF1
Gene Name
ELL associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033083.6 Intron NP_149074.3
XM_005265518.1 Intron XP_005265575.1
XM_011534165.1 Intron XP_011532467.1
XM_011534166.1 Intron XP_011532468.1
Gene
METTL6
Gene Name
methyltransferase like 6
There are no transcripts associated with this gene.

View Full Product Details