Product Details

SNP ID
rs2303040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38647968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTGAGGGGGCCCGGGGAGGGG[C/T]GGGAGGTCCTGAAAGGTAACTGGAG
Phenotype
MIM: 604638
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTN4 PubMed Links

Gene Details

Gene
ACTN4
Gene Name
actinin alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322033.1 Intron NP_001308962.1
NM_004924.5 Intron NP_004915.2
XM_005259281.4 Intron XP_005259338.1
XM_006723406.2 Intron XP_006723469.1
XM_017027331.1 Intron XP_016882820.1

View Full Product Details