Product Details
- SNP ID
-
rs2319707
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:168099415 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACCTTTCCTCACCTGTGGGCTGACT[A/G]CTGTGTGTTTAATCAAGAGGGCAAC
- Phenotype
-
MIM: 608008
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA10
PubMed Links
Gene Details
- Gene
- ANXA10
- Gene Name
- annexin A10
View Full Product Details