Product Details

SNP ID
hCV15763573
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48961490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCTGAGATGATGCCAGATTTCA[G/A]CCTGAGAGCAGAGTTTTGGAAGGTC
Phenotype
MIM: 300281 MIM: 300713
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
KCND1 PubMed Links

Gene Details

Gene
KCND1
Gene Name
potassium voltage-gated channel subfamily D member 1
There are no transcripts associated with this gene.

Gene
OTUD5
Gene Name
OTU deubiquitinase 5
There are no transcripts associated with this gene.

View Full Product Details