Product Details

SNP ID
rs3026401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31785976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAACTTCAAAACCCTATATGGTA[C/T]TCATTTCTTACACTAGATTTGCCTT
Phenotype
MIM: 606985 MIM: 607108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELP4 PubMed Links

Gene Details

Gene
ELP4
Gene Name
elongator acetyltransferase complex subunit 4
There are no transcripts associated with this gene.

Gene
PAX6
Gene Name
paired box 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000280.4 5545 UTR 3 NP_000271.1
NM_001127612.1 5545 UTR 3 NP_001121084.1
NM_001258462.1 5545 UTR 3 NP_001245391.1
NM_001258463.1 5545 UTR 3 NP_001245392.1
NM_001258464.1 5545 UTR 3 NP_001245393.1
NM_001258465.1 5545 UTR 3 NP_001245394.1
NM_001310158.1 5545 UTR 3 NP_001297087.1
NM_001310159.1 5545 Intron NP_001297088.1
NM_001310160.1 5545 UTR 3 NP_001297089.1
NM_001310161.1 5545 UTR 3 NP_001297090.1
NM_001604.5 5545 UTR 3 NP_001595.2

View Full Product Details