Product Details

SNP ID
rs3026636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37965395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTGATGATGTTTGTCACATCT[A/G]CTTCTTAACTGGATGGTTGATGTGT
Phenotype
MIM: 604414 MIM: 602229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6820 PubMed Links

Gene Details

Gene
MIR6820
Gene Name
microRNA 6820
There are no transcripts associated with this gene.

Gene
POLR2F
Gene Name
RNA polymerase II subunit F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301129.1 Intron NP_001288058.1
NM_001301130.1 Intron NP_001288059.1
NM_001301131.1 Intron NP_001288060.1
NM_021974.4 Intron NP_068809.1
XM_017028832.1 Intron XP_016884321.1
Gene
SOX10
Gene Name
SRY-box 10
There are no transcripts associated with this gene.

View Full Product Details