Product Details

SNP ID
rs2361177
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:136765773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCGGGGTGGCAGCGCCGGGGA[A/T]GGGGGCGCGCTGCCGGCGCAGACTT
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
THSD7B PubMed Links

Gene Details

Gene
THSD7B
Gene Name
thrombospondin type 1 domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316349.1 Intron NP_001303278.1
XM_017005049.1 Intron XP_016860538.1

View Full Product Details