Product Details

SNP ID
hCV15778012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54074944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTAGATTATTGAGGTGAAATT[C/T]GGCCGCGCCCTCTGTAGAATCAAGG
Phenotype
MIM: 616802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TARM1 PubMed Links

Gene Details

Gene
TARM1
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
There are no transcripts associated with this gene.

View Full Product Details