Product Details

SNP ID
rs3087226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22727230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCGAGCCCACCGGGAACGAAA[C/G]AGAAGCTCTATCTCCCCTCCAGGAG
Phenotype
MIM: 147620
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL6 PubMed Links

Gene Details

Gene
IL6
Gene Name
interleukin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000600.4 422 UTR 5 NP_000591.1
NM_001318095.1 422 UTR 5 NP_001305024.1
XM_005249745.4 422 UTR 5 XP_005249802.1
XM_011515390.2 422 UTR 5 XP_011513692.1
Gene
LOC541472
Gene Name
uncharacterized LOC541472
There are no transcripts associated with this gene.

View Full Product Details