Product Details

SNP ID
rs2412570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40847970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGTTCTCAGGGTACTCTCCTG[C/T]TTGTCAGCCAAGCCCCTGGGCTGGG
Phenotype
MIM: 605123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPINT1 PubMed Links

Gene Details

Gene
SPINT1
Gene Name
serine peptidase inhibitor, Kunitz type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032367.1 Intron NP_001027539.1
NM_003710.3 Intron NP_003701.1
NM_181642.2 Intron NP_857593.1
XM_006720657.1 Intron XP_006720720.1
XM_011521957.1 Intron XP_011520259.1
XM_011521958.1 Intron XP_011520260.1

View Full Product Details