Product Details

SNP ID
rs2467575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74863247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCCAGCCGCTGCAGTAGAGGC[A/C]TGAGAGGGGGTAACAAAAGGGGAAG
Phenotype
MIM: 103270 MIM: 138254
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FDXR PubMed Links

Gene Details

Gene
FDXR
Gene Name
ferredoxin reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258012.3 Intron NP_001244941.2
NM_001258013.3 Intron NP_001244942.2
NM_001258014.3 Intron NP_001244943.2
NM_001258015.2 Intron NP_001244944.1
NM_001258016.3 Intron NP_001244945.2
NM_004110.5 Intron NP_004101.3
NM_024417.4 Intron NP_077728.3
XM_006721772.1 Intron XP_006721835.1
XM_011524527.1 Intron XP_011522829.1
XM_011524528.1 Intron XP_011522830.1
XM_011524529.1 Intron XP_011522831.1
XM_011524531.2 Intron XP_011522833.1
XM_011524532.2 Intron XP_011522834.1
XM_011524533.2 Intron XP_011522835.1
Gene
GRIN2C
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
There are no transcripts associated with this gene.

View Full Product Details