Product Details

SNP ID
rs2442422
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4988650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATCTATATACTGGAGTGAGAGA[G/T]CAAGGGAAATCATTCAAATAAATTA
Phenotype
MIM: 605470
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MMP26 PubMed Links

Gene Details

Gene
MMP26
Gene Name
matrix metallopeptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021801.4 Intron NP_068573.2
XM_011520219.2 Intron XP_011518521.1

View Full Product Details