Product Details

SNP ID
rs2461831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119782951 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACCAGGTCCTTCTTCTCTCCAC[A/T]TTACAGCCCAGCTCGGCTGAAGCTA
Phenotype
MIM: 603065
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NR1I2 PubMed Links

Gene Details

Gene
NR1I2
Gene Name
nuclear receptor subfamily 1 group I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003889.3 Intron NP_003880.3
NM_022002.2 Intron NP_071285.1
NM_033013.2 Intron NP_148934.1

View Full Product Details