Product Details

SNP ID
rs2462707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7162280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTTCTTATTCTTTCTTTTTCT[A/C]ATTTTTGTACGATCTGGATCTTCCT
Phenotype
MIM: 615392
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SFMBT2 PubMed Links

Gene Details

Gene
SFMBT2
Gene Name
Scm-like with four mbt domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018039.1 4368 UTR 3 NP_001018049.1
NM_001029880.2 4368 UTR 3 NP_001025051.1
XM_005252551.2 4368 UTR 3 XP_005252608.1
XM_006717490.2 4368 UTR 3 XP_006717553.1
XM_011519611.1 4368 UTR 3 XP_011517913.1
XM_017016466.1 4368 UTR 3 XP_016871955.1

View Full Product Details