Product Details

SNP ID
rs2797581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93310323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTGAGAAAGGCTCTTTAACCTA[C/T]GACAGCTTCCTAAGATGCTGAACTT
Phenotype
MIM: 604603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOF PubMed Links

Gene Details

Gene
MYOF
Gene Name
myoferlin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013451.3 Intron NP_038479.1
NM_133337.2 Intron NP_579899.1
XM_005269693.4 Intron XP_005269750.1
XM_005269694.4 Intron XP_005269751.1
XM_011539632.2 Intron XP_011537934.1
XM_017016068.1 Intron XP_016871557.1
XM_017016069.1 Intron XP_016871558.1
XM_017016070.1 Intron XP_016871559.1

View Full Product Details