Product Details

SNP ID
rs4015581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103065557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAAGATATCACACTGGTCCATTA[C/T]ACTGATGGTATTATGCTGGTTAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf42 PubMed Links

Gene Details

Gene
C12orf42
Gene Name
chromosome 12 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099336.2 Intron NP_001092806.1
NM_001278419.1 Intron NP_001265348.1
NM_001278420.1 Intron NP_001265349.1
NM_198521.3 Intron NP_940923.2
XM_006719377.3 Intron XP_006719440.1
XM_011538291.1 Intron XP_011536593.1
XM_011538292.2 Intron XP_011536594.1
XM_011538293.2 Intron XP_011536595.1
XM_011538294.2 Intron XP_011536596.1
XM_011538295.1 Intron XP_011536597.1
XM_011538297.2 Intron XP_011536599.1
XM_011538300.1 Intron XP_011536602.1
XM_011538305.1 Intron XP_011536607.1
XM_011538306.1 Intron XP_011536608.1
XM_011538308.2 Intron XP_011536610.1
XM_011538312.2 Intron XP_011536614.1
XM_011538315.2 Intron XP_011536617.1
XM_017019276.1 Intron XP_016874765.1
XM_017019277.1 Intron XP_016874766.1
XM_017019278.1 Intron XP_016874767.1
XM_017019279.1 Intron XP_016874768.1
XM_017019280.1 Intron XP_016874769.1
XM_017019281.1 Intron XP_016874770.1

View Full Product Details