Product Details

SNP ID
rs2840050
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:85066288 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAGGCACAACATTGATTTTAGGA[A/G]TAGACGAATTATCTCTTCTGCTACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR63 PubMed Links

Gene Details

Gene
WDR63
Gene Name
WD repeat domain 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288563.1 Intron NP_001275492.1
NM_145172.4 Intron NP_660155.2

View Full Product Details