Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001112726.2 | Intron | NP_001106197.1 | |||
NM_015005.2 | Intron | NP_055820.2 | |||
XM_005267550.3 | Intron | XP_005267607.1 | |||
XM_011536665.2 | Intron | XP_011534967.1 | |||
XM_011536666.2 | Intron | XP_011534968.1 | |||
XM_011536667.2 | Intron | XP_011534969.1 | |||
XM_017021227.1 | Intron | XP_016876716.1 | |||
XM_017021228.1 | Intron | XP_016876717.1 | |||
XM_017021229.1 | Intron | XP_016876718.1 | |||
XM_017021230.1 | Intron | XP_016876719.1 |