Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001160.2 | 408 | UTR 5 | NP_001151.1 | ||
NM_013229.2 | 408 | UTR 5 | NP_037361.1 | ||
NM_181861.1 | 408 | UTR 5 | NP_863651.1 | ||
NM_181868.1 | 408 | UTR 5 | NP_863658.1 | ||
NM_181869.1 | 408 | UTR 5 | NP_863659.1 | ||
XM_017019250.1 | 408 | UTR 5 | XP_016874739.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_153687.3 | 408 | Intron | NP_710154.1 | ||
NM_201612.2 | 408 | Intron | NP_963906.1 | ||
NM_201613.2 | 408 | Intron | NP_963907.1 |