Product Details

SNP ID
hCV15822634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112059136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTATCCTGAGCACTCCTATTTCT[G/T]GCTACAATGCAGTTCTTTCCCTTCA
Phenotype
MIM: 608770
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DLAT PubMed Links

Gene Details

Gene
DLAT
Gene Name
dihydrolipoamide S-acetyltransferase
There are no transcripts associated with this gene.

Gene
PIH1D2
Gene Name
PIH1 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details