Product Details

SNP ID
rs2853208
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3660176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTACCTGCAGGAGGCTGTGCTC[C/T]GGGTCTCCCTGGGGGTTCAGCTGGT
Phenotype
MIM: 607114 MIM: 603130
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAM33 PubMed Links

Gene Details

Gene
ADAM33
Gene Name
ADAM metallopeptidase domain 33
There are no transcripts associated with this gene.

Gene
ATRN
Gene Name
attractin
There are no transcripts associated with this gene.

Gene
GFRA4
Gene Name
GDNF family receptor alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022139.3 711 Silent Mutation CCA,CCG P237P NP_071422.1
NM_145762.2 711 Silent Mutation CCA,CCG P267P NP_665705.1
XM_005260793.2 711 Intron XP_005260850.1

View Full Product Details