Product Details

SNP ID
rs2853747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87976077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGAAAATAGTAAAATTAAATTC[A/C]CCTTTGGAATAATTATACCTATATA
Phenotype
MIM: 166490
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPP1 PubMed Links

Gene Details

Gene
SPP1
Gene Name
secreted phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000582.2 Intron NP_000573.1
NM_001040058.1 Intron NP_001035147.1
NM_001040060.1 Intron NP_001035149.1
NM_001251829.1 Intron NP_001238758.1
NM_001251830.1 Intron NP_001238759.1
XM_017008564.1 Intron XP_016864053.1

View Full Product Details