Product Details

SNP ID
rs2148342
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:41551784 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAATGATGGCGACTTGAAGGTAGG[A/G]TGGCTTAGGGAATGGAGCCAGGGAC
Phenotype
MIM: 608924
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXP4 PubMed Links

Gene Details

Gene
FOXP4
Gene Name
forkhead box P4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012426.1 Intron NP_001012426.1
NM_001012427.1 Intron NP_001012427.1
NM_138457.2 Intron NP_612466.1
XM_006714991.3 Intron XP_006715054.1
XM_011514289.2 Intron XP_011512591.1
XM_011514290.2 Intron XP_011512592.1
XM_011514291.2 Intron XP_011512593.1
XM_011514292.2 Intron XP_011512594.1
XM_011514293.2 Intron XP_011512595.1
XM_017010233.1 Intron XP_016865722.1
XM_017010234.1 Intron XP_016865723.1
Gene
FOXP4-AS1
Gene Name
FOXP4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details