Product Details
- SNP ID
-
rs2838950
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:45506383 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGCTGTCCCGTGGCTCTGCACCC[C/T]GCGTTATAAACACCAAGGTGGGATG
- Phenotype
-
MIM: 120328
MIM: 600424
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COL18A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112798876] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL18A1
- Gene Name
- collagen type XVIII alpha 1 chain
- Gene
- SLC19A1
- Gene Name
- solute carrier family 19 member 1
View Full Product Details