Product Details
- SNP ID
-
rs2166447
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:208169199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAGGGTCCATTGATGAGGCTTGC[A/G]GGGGGTGTCTATAAACCCCTGTTTC
- Phenotype
-
MIM: 615536
MIM: 123660
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf80
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5838108] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf80
- Gene Name
- chromosome 2 open reading frame 80
- Gene
- CRYGA
- Gene Name
- crystallin gamma A
There are no transcripts associated with this gene.
View Full Product Details