Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001174087.1 | 6959 | UTR 3 | NP_001167558.1 | ||
NM_001174088.1 | 6959 | UTR 3 | NP_001167559.1 | ||
NM_006534.3 | 6959 | UTR 3 | NP_006525.2 | ||
NM_181659.2 | 6959 | UTR 3 | NP_858045.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001161841.1 | 6959 | Intron | NP_001155313.1 | ||
NM_018837.3 | 6959 | Intron | NP_061325.1 | ||
NM_198596.2 | 6959 | Intron | NP_940998.2 | ||
XM_005260458.2 | 6959 | UTR 3 | XP_005260515.1 | ||
XM_006723827.1 | 6959 | Intron | XP_006723890.1 | ||
XM_006723828.1 | 6959 | Intron | XP_006723891.1 | ||
XM_006723829.2 | 6959 | Intron | XP_006723892.1 | ||
XM_006723830.1 | 6959 | Intron | XP_006723893.1 | ||
XM_011528913.1 | 6959 | Intron | XP_011527215.1 | ||
XM_011528914.1 | 6959 | Intron | XP_011527216.1 | ||
XM_011528915.1 | 6959 | Intron | XP_011527217.1 | ||
XM_011528916.1 | 6959 | Intron | XP_011527218.1 | ||
XM_017027955.1 | 6959 | UTR 3 | XP_016883444.1 |