Product Details

SNP ID
rs2913916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176546849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCATTGTAATAGTCGCATCGAG[T/C]GCTACCCAGTGTCTGCAGCATCCTT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CDHR2 PubMed Links

Gene Details

Gene
CDHR2
Gene Name
cadherin related family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171976.1 Intron NP_001165447.1
NM_017675.4 Intron NP_060145.3
Gene
RNF44
Gene Name
ring finger protein 44
There are no transcripts associated with this gene.

View Full Product Details