Product Details

SNP ID
rs2231163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88094735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTCCAAAGAGTTATCACAATCA[C/T]GCCCTCTGAGTTTAATCTTTATTGT
Phenotype
MIM: 603756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCG2 PubMed Links

Gene Details

Gene
ABCG2
Gene Name
ATP binding cassette subfamily G member 2 (Junior blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257386.1 Intron NP_001244315.1
NM_004827.2 Intron NP_004818.2
XM_005263354.3 Intron XP_005263411.1
XM_005263355.3 Intron XP_005263412.1
XM_005263356.3 Intron XP_005263413.1
XM_011532420.2 Intron XP_011530722.1
XM_017008852.1 Intron XP_016864341.1
XM_017008853.1 Intron XP_016864342.1

View Full Product Details