Product Details

SNP ID
rs2231345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68866156 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCAGAAAACACGATGAGCAAAGG[G/T]TTATGCTTATTCTGGGCACTGGTAT
Phenotype
MIM: 608748
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BMP10 PubMed Links

Gene Details

Gene
BMP10
Gene Name
bone morphogenetic protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014482.1 909 Missense Mutation NP_055297.1

View Full Product Details