Product Details

SNP ID
rs2906879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15893309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCTCACACAACCCCCAGACCTC[A/T]CACAACGCCCAGATGTTTGGACCAC
Phenotype
MIM: 604426
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP4F2 PubMed Links

Gene Details

Gene
CYP4F2
Gene Name
cytochrome P450 family 4 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082.4 Intron NP_001073.3

View Full Product Details