Product Details

SNP ID
rs2231571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36115242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTACCCTTCGCGGACTGGGCGA[A/G]CTCTTGCCACTCCTACCTCCGGGCT
Phenotype
MIM: 616442 MIM: 180662 MIM: 601303 MIM: 613583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OVOL3 PubMed Links

Gene Details

Gene
OVOL3
Gene Name
ovo like zinc finger 3
There are no transcripts associated with this gene.

Gene
POLR2I
Gene Name
RNA polymerase II subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006233.4 257 UTR 5 NP_006224.1
Gene
TBCB
Gene Name
tubulin folding cofactor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281.2 257 UTR 5 NP_001272.2
NM_001300971.1 257 Intron NP_001287900.1
Gene
WDR62
Gene Name
WD repeat domain 62
There are no transcripts associated with this gene.

View Full Product Details