Product Details

SNP ID
rs2231882
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:27614638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATGTGGACTCACAACACCGTG[A/G]CCTGTTTGAGGGGTGAGGGACCATT
Phenotype
MIM: 164940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGR PubMed Links

Gene Details

Gene
FGR
Gene Name
FGR proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042729.1 Intron NP_001036194.1
NM_001042747.1 Intron NP_001036212.1
NM_005248.2 Intron NP_005239.1
XM_006710452.2 Intron XP_006710515.1
XM_011541010.1 Intron XP_011539312.1
XM_011541011.2 Intron XP_011539313.1
XM_011541012.1 Intron XP_011539314.1
XM_011541013.2 Intron XP_011539315.1
XM_011541014.2 Intron XP_011539316.1
XM_017000673.1 Intron XP_016856162.1
XM_017000674.1 Intron XP_016856163.1

View Full Product Details