Product Details

SNP ID
rs2232198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:142004959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAGCATCCATGACAGTGCCCAC[C/T]CCCACCGTCAAGGCCATGGTGGGCA
Phenotype
MIM: 601798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNPDA1 PubMed Links

Gene Details

Gene
GNPDA1
Gene Name
glucosamine-6-phosphate deaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005471.4 634 Silent Mutation GGA,GGG G189G NP_005462.1
XM_005268348.1 634 Silent Mutation GGA,GGG G218G XP_005268405.1
XM_006714747.2 634 Silent Mutation GGA,GGG G189G XP_006714810.1
XM_011537537.1 634 Silent Mutation GGA,GGG G189G XP_011535839.1

View Full Product Details