Product Details

SNP ID
rs2068136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:129095038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGTGGTATTTACATAAAGTGAA[C/T]ATCTCAGGTGAAAACTTTTATGTCT
Phenotype
MIM: 611399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4orf33 PubMed Links

Gene Details

Gene
C4orf33
Gene Name
chromosome 4 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099783.1 Intron NP_001093253.1
NM_173487.2 Intron NP_775758.2
XM_005262734.4 Intron XP_005262791.1
XM_017007722.1 Intron XP_016863211.1
XM_017007723.1 Intron XP_016863212.1
XM_017007724.1 Intron XP_016863213.1
Gene
SCLT1
Gene Name
sodium channel and clathrin linker 1
There are no transcripts associated with this gene.

View Full Product Details