Product Details

SNP ID
rs2070441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23402290 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTACCCCATCCACCTCTTTTAGGC[C/T]CCCCCTAACTTGCCTGTTTCCCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107985555 PubMed Links

Gene Details

Gene
LOC107985555
Gene Name
coiled-coil domain-containing protein 188-like
There are no transcripts associated with this gene.

Gene
ZDHHC8P1
Gene Name
zinc finger DHHC-type containing 8 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details