Product Details

SNP ID
rs2070843
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:150444449 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGGCCTAACCAATCAGCAACAGAT[A/C]CTGCTGCTCCCCAAATGACTCCCCA
Phenotype
MIM: 130620
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPS14 PubMed Links

Gene Details

Gene
RPS14
Gene Name
ribosomal protein S14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025070.1 Intron NP_001020241.1
NM_001025071.1 Intron NP_001020242.1
NM_005617.3 Intron NP_005608.1
XM_017009684.1 Intron XP_016865173.1

View Full Product Details