Product Details

SNP ID
rs2071044
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:52813585 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCTGGGGACAGGAACATGGAGGG[T/C]AGGGAGTCCTGGCGTCCCTTTAGAA
Phenotype
MIM: 146650 MIM: 600564
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
ITIH3 PubMed Links

Gene Details

Gene
ITIH3
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
There are no transcripts associated with this gene.

Gene
ITIH4
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166449.1 Intron NP_001159921.1
NM_002218.4 Intron NP_002209.2

View Full Product Details