Product Details

SNP ID
rs2223665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:297874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCCCACCGGCTGGCCGCGGGGA[C/T]CCGAAGGGCCGTCGGAGAGATCCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
There are no transcripts associated with this gene.

Gene
ZCCHC3
Gene Name
zinc finger CCHC-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033089.6 312 Silent Mutation GAC,GAT D96D NP_149080.2

View Full Product Details